![9-Vinylanthracene 97%](/deepweb/assets/sigmaaldrich/product/structures/401/597/85a01d1d-1529-4bc8-8706-050b4c9a868d/640/85a01d1d-1529-4bc8-8706-050b4c9a868d.png)
9-Vinylanthracene
![9-Vinylanthracene 97%](/deepweb/assets/sigmaaldrich/product/structures/401/597/85a01d1d-1529-4bc8-8706-050b4c9a868d/640/85a01d1d-1529-4bc8-8706-050b4c9a868d.png)
Synonym(s):
9-Vinylanthracene
Empirical Formula (Hill Notation):
C16H12
CAS Number:
Molecular Weight:
204.27
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing