Skip to Content
Merck
9-Undecynoic acid 90%

9-Undecynoic acid

9-Undecynoic acid 90%
Empirical Formula (Hill Notation):
C11H18O2
CAS Number:
Molecular Weight:
182.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing