Skip to Content
MilliporeSigma
7-Bromo-6-chloro-7-deazapurine 95%

7-Bromo-6-chloro-7-deazapurine

7-Bromo-6-chloro-7-deazapurine 95%
Synonym(s):
5-Bromo-4-chloro-7H-pyrrolo[2,3-d]pyrimidine
Empirical Formula (Hill Notation):
C6H3BrClN3
CAS Number:
Molecular Weight:
232.47

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing