Skip to Content
Merck
6-Chloro-4-chromanone 97%

6-Chloro-4-chromanone

6-Chloro-4-chromanone 97%
Empirical Formula (Hill Notation):
C9H7ClO2
CAS Number:
Molecular Weight:
182.60
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing