
6-Benzoyl-2-naphthol

Synonym(s):
6-Hydroxy-2-naphthyl phenyl ketone
Linear Formula:
C6H5COC10H6OH
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing