6-Aminopyridine-2-carboxylic acid
Synonym(s):
6-Amino-2-picolinic acid
Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing