Skip to Content
Merck
6-Amino-2,3-dimethylquinoxaline 95%

6-Amino-2,3-dimethylquinoxaline

6-Amino-2,3-dimethylquinoxaline 95%
Synonym(s):
2,3-Dimethylquinoxalin-6-amine
Empirical Formula (Hill Notation):
C10H11N3
CAS Number:
Molecular Weight:
173.21

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing