
6,7-Dimethoxy-4-methylcoumarin

Empirical Formula (Hill Notation):
C12H12O4
CAS Number:
Molecular Weight:
220.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing