Skip to Content
Merck
6-[(4-Methyl-1-piperazinyl)methyl]nicotinic acid AldrichCPR

6-[(4-Methyl-1-piperazinyl)methyl]nicotinic acid

6-[(4-Methyl-1-piperazinyl)methyl]nicotinic acid AldrichCPR
Empirical Formula (Hill Notation):
C12H17N3O2
Molecular Weight:
235.28

Products

Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing