6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde
Synonym(s):
6-(4-Fluorophenyl)pyridine-2-carbaldehyde
Empirical Formula (Hill Notation):
C12H8FNO
CAS Number:
Molecular Weight:
201.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing