Skip to Content
Merck
5-(p-Methylphenyl)-5-phenylhydantoin ≥99%

5-(p-Methylphenyl)-5-phenylhydantoin

5-(p-Methylphenyl)-5-phenylhydantoin ≥99%
Synonym(s):
5-(4-Methylphenyl)-,5-phenylimidazolidine-2,4-dione, 5-Phenyl-5-(p-tolyl)hydantoin, MPPH, NSC 32105
Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing