Skip to Content
Merck
5-Methyl-2-furoic acid 97%

5-Methyl-2-furoic acid

5-Methyl-2-furoic acid 97%
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing