Skip to Content
Merck
5-Hydroxy-1-methyl-1H-pyrazole 97%

5-Hydroxy-1-methyl-1H-pyrazole

5-Hydroxy-1-methyl-1H-pyrazole 97%
Synonym(s):
1-Methyl-1H-pyrazol-5-ol, 1-Methyl-5-hydroxy-1H-pyrazole, 1-Methyl-5-hydroxypyrazole, 1-Methylpyrazol-5-ol, 2-Methyl-2H-pyrazol-3-ol, 5-Hydroxy-1-methylpyrazole
Empirical Formula (Hill Notation):
C4H6N2O
CAS Number:
Molecular Weight:
98.10

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing