Skip to Content
Merck
5-Fluoro-2-nitrobenzoic acid 98%

5-Fluoro-2-nitrobenzoic acid

5-Fluoro-2-nitrobenzoic acid 98%
Linear Formula:
FC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
185.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing