Skip to Content
Merck
5-Fluoro-2-hydroxypyrimidine 97%

5-Fluoro-2-hydroxypyrimidine

5-Fluoro-2-hydroxypyrimidine 97%
Synonym(s):
5-Fluoro-2(1H)-pyrimidone
Empirical Formula (Hill Notation):
C4H3FN2O
CAS Number:
Molecular Weight:
114.08

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing