Skip to Content
Merck
5-Chloro-1-phenyl-1H-tetrazole 97%

5-Chloro-1-phenyl-1H-tetrazole

5-Chloro-1-phenyl-1H-tetrazole 97%
Empirical Formula (Hill Notation):
C7H5ClN4
CAS Number:
Molecular Weight:
180.59
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing