Skip to Content
Merck
5-Bromoisatoic anhydride 97%

5-Bromoisatoic anhydride

5-Bromoisatoic anhydride 97%
Synonym(s):
6-Bromo-1,2-dihydro-4H-3,1-benzoxazine-2,4-dione
Empirical Formula (Hill Notation):
C8H4BrNO3
CAS Number:
Molecular Weight:
242.03

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing