Skip to Content
Merck
5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt ~97% (HPLC)

5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt

5-Bromo-6-chloro-3-indolyl phosphate p-toluidine salt ~97% (HPLC)
Synonym(s):
Magenta phosphate
Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing