Skip to Content
Merck
5-Bromo-3-nitrosalicylaldehyde 97%

5-Bromo-3-nitrosalicylaldehyde

5-Bromo-3-nitrosalicylaldehyde 97%
Linear Formula:
BrC6H2(NO2)-2-(OH)CHO
CAS Number:
Molecular Weight:
246.01

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing