Skip to Content
Merck
5-Bromo-2,4-dimethoxypyrimidine 97%

5-Bromo-2,4-dimethoxypyrimidine

5-Bromo-2,4-dimethoxypyrimidine 97%
Empirical Formula (Hill Notation):
C6H7BrN2O2
CAS Number:
Molecular Weight:
219.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing