Skip to Content
Merck
5,6-Dihydro-4-methoxy-2H-pyran 95%

5,6-Dihydro-4-methoxy-2H-pyran

5,6-Dihydro-4-methoxy-2H-pyran 95%
Synonym(s):
4-Methoxy-5,6-dihydropyran
Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing