Skip to Content
Merck
5-(5-Bromo-2-thienyl)-1H-tetrazole solution 30% in DMF

5-(5-Bromo-2-thienyl)-1H-tetrazole solution

5-(5-Bromo-2-thienyl)-1H-tetrazole solution 30% in DMF
Empirical Formula (Hill Notation):
C5H3BrN4S
CAS Number:
Molecular Weight:
231.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing