Skip to Content
Merck
(4R,5S)-(−)-1,5-Dimethyl-4-phenyl-2-imidazolidinone 97%

(4R,5S)-(−)-1,5-Dimethyl-4-phenyl-2-imidazolidinone

(4R,5S)-(−)-1,5-Dimethyl-4-phenyl-2-imidazolidinone 97%
Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing