Skip to Content
Merck
4-Nitrothioanisole 96%

4-Nitrothioanisole

4-Nitrothioanisole 96%
Synonym(s):
Methyl-4-nitrophenyl sulfide
Linear Formula:
O2NC6H4SCH3
CAS Number:
Molecular Weight:
169.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing