4-Nitrophenyl α-D-maltopentaoside
Synonym(s):
4-Nitrophenyl α-D-penta-(1→4)-glucopyranoside
Empirical Formula (Hill Notation):
C36H55NO28
CAS Number:
Molecular Weight:
949.81
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing