Skip to Content
Merck
4-Methoxy-TEMPO 97%

4-Methoxy-TEMPO

4-Methoxy-TEMPO 97%
Synonym(s):
4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
Empirical Formula (Hill Notation):
C10H20NO2
CAS Number:
Molecular Weight:
186.27

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing