Skip to Content
Merck
4-Iodo-2-nitroaniline 97%

4-Iodo-2-nitroaniline

4-Iodo-2-nitroaniline 97%
Synonym(s):
4-Iodo-2-nitrophenylamine
Empirical Formula (Hill Notation):
C6H5IN2O2
CAS Number:
Molecular Weight:
264.02

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing