Skip to Content
Merck
4-(Hydroxymethyl)phenoxyacetic acid ≥97.0%

4-(Hydroxymethyl)phenoxyacetic acid

4-(Hydroxymethyl)phenoxyacetic acid ≥97.0%
Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing