Skip to Content
Merck
4-Hydroxy-D-phenylglycine ≥98%

4-Hydroxy-D-phenylglycine

4-Hydroxy-D-phenylglycine ≥98%
Synonym(s):
(2R)-2-Amino-2-(4-hydroxyphenyl)acetic acid
Linear Formula:
HOC6H4CH(NH2)CO2H
CAS Number:
Molecular Weight:
167.16
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing