Skip to Content
Merck
4-Hydroxy-6-(trifluoromethyl)pyrimidine 99%

4-Hydroxy-6-(trifluoromethyl)pyrimidine

4-Hydroxy-6-(trifluoromethyl)pyrimidine 99%
Synonym(s):
6-(Trifluoromethyl)-4-pyrimidinol
Empirical Formula (Hill Notation):
C5H3F3N2O
CAS Number:
Molecular Weight:
164.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing