![4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/195/962/fce02118-5198-4a53-93e5-830e761a38a1/640/fce02118-5198-4a53-93e5-830e761a38a1.png)
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid
![4H-Thieno[3,2-b]pyrrole-5-carboxylic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/195/962/fce02118-5198-4a53-93e5-830e761a38a1/640/fce02118-5198-4a53-93e5-830e761a38a1.png)
Empirical Formula (Hill Notation):
C7H5NO2S
Molecular Weight:
167.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing