Skip to Content
Merck
4-Fluororesorcinol 97%

4-Fluororesorcinol

4-Fluororesorcinol 97%
Synonym(s):
4-Fluoro-1,3-benzenediol, 4-Fluoro-1,3-dihydroxybenzene
Empirical Formula (Hill Notation):
C6H5FO2
CAS Number:
Molecular Weight:
128.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing