Skip to Content
Merck
4-Fluoro-2-nitroaniline 97%

4-Fluoro-2-nitroaniline

4-Fluoro-2-nitroaniline 97%
Linear Formula:
FC6H3(NO2)NH2
CAS Number:
Molecular Weight:
156.11
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing