Skip to Content
Merck
4-Bromo-2-fluorobenzyl bromide 98%

4-Bromo-2-fluorobenzyl bromide

4-Bromo-2-fluorobenzyl bromide 98%
Synonym(s):
4,α-Dibromo-2-fluorotoluene
Linear Formula:
BrC6H3(F)CH2Br
CAS Number:
Molecular Weight:
267.92
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing