Skip to Content
Merck
4-Benzoylbenzoic acid N-succinimidyl ester 97%

4-Benzoylbenzoic acid N-succinimidyl ester

4-Benzoylbenzoic acid N-succinimidyl ester 97%
Synonym(s):
4-Benzoylbenzoic acid, succinimidyl ester
Empirical Formula (Hill Notation):
C18H13NO5
CAS Number:
Molecular Weight:
323.30

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing