Skip to Content
Merck
4,5-Diphenyl-1,3-dioxol-2-one 99%

4,5-Diphenyl-1,3-dioxol-2-one

4,5-Diphenyl-1,3-dioxol-2-one 99%
Synonym(s):
1,2-Diphenylvinylene carbonate
Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing