![4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/336/452/a3630b1b-df3e-44ad-8b07-2521d4dfc53b/640/a3630b1b-df3e-44ad-8b07-2521d4dfc53b.png)
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid
![4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/336/452/a3630b1b-df3e-44ad-8b07-2521d4dfc53b/640/a3630b1b-df3e-44ad-8b07-2521d4dfc53b.png)
Synonym(s):
4,5,6,7-Tetrahydrobenzothiophene-3-carboxylic acid
Empirical Formula (Hill Notation):
C9H10O2S
CAS Number:
Molecular Weight:
182.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing