
4,4′-Dihydroxydiphenyl sulfone bisfluorosulfinate

Empirical Formula (Hill Notation):
C12H8F2O8S3
CAS Number:
Molecular Weight:
414.38
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing