Skip to Content
Merck
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate ≥98.0% (TLC)

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate ≥98.0% (TLC)
Synonym(s):
meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulfonic acid tetrasodium salt
Empirical Formula (Hill Notation):
C44H26N4Na4O12S4 · xH2O
CAS Number:
Molecular Weight:
1022.92 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Special Grade