Skip to Content
Merck
4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 monoethoxylate solution 10 μg/mL in acetone, analytical standard

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 monoethoxylate solution

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 monoethoxylate solution 10 μg/mL in acetone, analytical standard
Synonym(s):
2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy-3,5-d2]ethanol, 2-[4-(3,6-Dimethyl-3-heptyl)phenoxy-3,5-d2]ethanol, 3,6,3-Nonylphenol-monoethoxylate, 363-NP1EO-D2, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-3,5-d2-monoethoxylate
Empirical Formula (Hill Notation):
C17D2H26O2
CAS Number:
Molecular Weight:
266.42
EC Number:

Products

Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique