Skip to Content
Merck
4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 diethoxylate solution 1 μg/mL in acetone, analytical standard

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 diethoxylate solution

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 diethoxylate solution 1 μg/mL in acetone, analytical standard
Synonym(s):
2-{2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy-3,5-d2]ethoxy}ethanol, 2-{2-[4-(3,6-Dimethyl-3-heptyl)phenoxy-3,5-d2]ethoxy}ethanol, 3,6,3-Nonylphenol diethoxylate-d2 (ring-3,5-d2), 363-NP2EO-D2
Empirical Formula (Hill Notation):
C19D2H30O3
CAS Number:
Molecular Weight:
310.47
EC Number:

Products

Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1