Skip to Content
Merck
4-(2-Pyridyl)benzaldehyde 99%

4-(2-Pyridyl)benzaldehyde

4-(2-Pyridyl)benzaldehyde 99%
Empirical Formula (Hill Notation):
C12H9NO
CAS Number:
Molecular Weight:
183.21

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing