Skip to Content
MilliporeSigma
3-Thiophenemalonic acid 97%

3-Thiophenemalonic acid

3-Thiophenemalonic acid 97%
Synonym(s):
α-Carboxythiophene-3-acetic acid, (3-Thienyl)malonic acid
Empirical Formula (Hill Notation):
C7H6O4S
CAS Number:
Molecular Weight:
186.19
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing