Skip to Content
Merck
3-Quinolineboronic acid technical grade

3-Quinolineboronic acid

3-Quinolineboronic acid technical grade
Synonym(s):
(3-Quinolinyl)boronic acid, (3-Quinolyl)boronic acid, Quinolin-3-ylboronic acid
Empirical Formula (Hill Notation):
C9H8BNO2
CAS Number:
Molecular Weight:
172.98

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing