Skip to Content
Merck
3-Phenylisoxazole-5-carboxylic acid 97%

3-Phenylisoxazole-5-carboxylic acid

3-Phenylisoxazole-5-carboxylic acid 97%
Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing