Skip to Content
Merck
3-Ethynyl-α,α,α-trifluorotoluene 97%

3-Ethynyl-α,α,α-trifluorotoluene

3-Ethynyl-α,α,α-trifluorotoluene 97%
Linear Formula:
HC≡CC6H4CF3
CAS Number:
Molecular Weight:
170.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing