Skip to Content
Merck
3-Ethyl-2-methylbenzothiazolium iodide

3-Ethyl-2-methylbenzothiazolium iodide

3-Ethyl-2-methylbenzothiazolium iodide
Empirical Formula (Hill Notation):
C10H12INS
CAS Number:
Molecular Weight:
305.18
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing