Skip to Content
Merck
3-Chloro-4-fluorophenylboronic acid ≥95%

3-Chloro-4-fluorophenylboronic acid

3-Chloro-4-fluorophenylboronic acid ≥95%
Synonym(s):
(3-Chloro-4-fluorophenyl-1-yl)boronic acid, 3-Chloro-4-fluorobenzeneboronic acid, 4-Fluoro-3-chlorophenylboronic acid
Linear Formula:
ClC6H3(F)B(OH)2
CAS Number:
Molecular Weight:
174.37

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing