Skip to Content
Merck
3-Chloro-2,4(3H,5H)-furandione ≥98.0% (HPLC)

3-Chloro-2,4(3H,5H)-furandione

3-Chloro-2,4(3H,5H)-furandione ≥98.0% (HPLC)
Synonym(s):
3-Chlorotetronic acid
Empirical Formula (Hill Notation):
C4H3ClO3
CAS Number:
Molecular Weight:
134.52

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing