Skip to Content
Merck
3-Bromoisoquinoline 95%

3-Bromoisoquinoline

3-Bromoisoquinoline 95%
Empirical Formula (Hill Notation):
C9H6BrN
CAS Number:
Molecular Weight:
208.05

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing